missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNFSF4 Monoclonal Antibody (OTI5D8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA806677

This item is not returnable.
View return policy
Description
The tumor necrosis factor superfamily member TNFSF4 is a type II membrane bound, non-covalently linked homotrimeric protein. It is expressed on antigen presenting cells, such as dendritic cells and activated B-cells, and also on other cells such as vascular endothelial cells, mast cells, and natural killer cells. TNFSF4 signals specifically through the TNFRSF4 receptor, is expressed predomitly on CD4+T cells but also on certain activated CD8+T cells. TNFRSF4/TNFSF4 functions as a costimulatory signal, which is required for a productive interaction between antigen presenting cells and their target T-cells. It enhances cell proliferation and survival, and increases expression of RANTES, IL-2, IL-3, and IFNgamma. TNFRSF4/TNFSF4 signaling plays an important role in immunotolerance.Specifications
TNFSF4 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P23510 | |
TNFSF4 | |
Full length human recombit protein of human TNFSF4 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
OTI5D8 | |
Unconjugated | |
TNFSF4 | |
CD134L, CD252, GP34, OX-40L, OX4OL, TNLG2B, TXGP1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
7292 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
TNFSF4 Monoclonal Antibody (OTI5D8), TrueMAB™, OriGene
Spot an opportunity for improvement?Share a Content Correction