missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTW™ Total Organic Carbon Test Kits
Test materials for the AQA of photometric routine and water analysis for multi-level result certainty of photoLab™, pHotoFlex™ and test kits
Brand: WTW™ 252037
View more versions of this product
This item is not returnable.
View return policy
Description
Specifications
SO4-1 TP: barium sulfate, turbidometric | |
PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral photometers | |
90 | |
25 Tests | |
CombiCheck Sulfate (SO4) cell test kit |
For measurement of TOC (Total organic carbon) | |
Certificates | |
50 to 800 mg/L | |
Photometric |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- TOC Cell Test Method: photometric 50 - 800 mg/l Spectroquant
Signal Word
- Warning
Hazard Category
- Substances and mixtures corrosive to metals Category 1
Hazard Statement
- H290-May be corrosive to metals.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
- MIXTURE LIST-Contains : sulphuric acid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction