missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTW™ Total Organic Carbon Test Kits
Test materials for the AQA of photometric routine and water analysis for multi-level result certainty of photoLab™, pHotoFlex™ and test kits
388.00 EUR - 415.00 EUR
Specifications
For Use With (Application) | For measurement of TOC (Total organic carbon) |
---|---|
For Use With (Equipment) | PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral photometers |
Includes | Certificates |
Sufficient For | 25 Tests |
Test Method | Photometric |
Product Code | Brand | Range | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Range | Price | Quantity & Availability | |||||
11789513
|
WTW™
252037 |
50 to 800 mg/L |
415.00 EUR
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11769513
|
WTW™
252036 |
5 to 80 mg/L |
388.00 EUR
75g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Specifications
For measurement of TOC (Total organic carbon) | |
Certificates | |
Photometric |
PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral photometers | |
25 Tests |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- TOC Cell Test Method: photometric 50 - 800 mg/l Spectroquant
Signal Word
- Warning
Hazard Category
- Substances and mixtures corrosive to metals Category 1
Hazard Statement
- H290-May be corrosive to metals.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
- MIXTURE LIST-Contains : sulphuric acid