Learn More
Invitrogen™ TRA-1-60 Alexa Fluor™ 488 Conjugate Kit for Live Cell Imaging
Molecular Probes™ human pluripotent stem cell (PSC) live imaging kits provide reagents for superior imaging of live human PSCs.
Brand:  Invitrogen™ A25618
442.70 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Product Code. 15564169
  • 471.00 EUR / 50 tests
Description
This TRA-1-60 Alexa Fluor™ 488 Conjugate Kit includes an optimally labeled primary antibody paired with reduced-background imaging medium (FluoroBrite™ DMEM) for the specific detection of the PSC marker TRA-1-60. For fixed cell staining of PSC markers and germ layer markers, we recommend the Pluripotent Stem Cell 4-Marker Immunocytochemistry Kit.
With the TRA-1-60 Alexa Fluor™ 488 Conjugate Kit for Live Cell Imaging, you can:
• Confirm the expression of a key pluripotent stem cell marker using an optimally dye-conjugated antibody
• Preserve cell health and maximize fluorescence signal with FluoroBrite™ DMEM
• Stain live cell samples with confidence using reagents that are tested to be free of common contaminants
Confirm Expression of a Key PSC Marker
Powered by the high performance of bright, photostable Alexa Fluor™ dyes, this kit enables reliable live-cell detection of the surface PSC marker TRA-1-60. This antibody conjugate has been demonstrated to show superior staining results when used with FluoroBrite™ DMEM.
Preserve Cell Health and Maximize Signal
Staining cells in FluoroBrite™ DMEM maximizes cell health and enhances signal-to-noise ratio when performing live-cell fluorescence imaging.
Stain Live-cell Samples with Confidence
Unlike most other conjugated antibodies, the reagents provided in this kit are 0.2 μm sterile-filtered and tested to be free of common contaminants.
What You Get
Easily scalable to any culture plate, dish, or slide format, the TRA-1-60 Alexa Fluor™ 488 Conjugate Kit comes with reagents sufficient to stain 50 samples using a 200 μL staining volume and includes:
• 200 μL of Alexa Fluor™ 488 Mouse Anti-Human TRA-1-60 (50X concentration)
• 500 mL FluoroBrite™ DMEM
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Alexa Fluor 488
  • 200 μL of Alexa Fluor™ 488 Mouse Anti-Human TRA-1-60 (50 x concentration) (store at 2°C to 8°C)
  • 500 mL FluoroBrite™ DMEM
  • Store at 2°C to 8°C, protect from light
)
Conjugate Kit
50 tests
Alexa Fluor™
SDS
Documents Certificates
Special Offers
Special Offers
  • Multiple Manufacturers
    Advance your proteomics research with essential tools
    Promo Code: 25977
    Expires2025-05-31
    Days
    54
    Hours
    11
    Minutes
    53
    Seconds
    58
Product Content Correction
Product Title
Invitrogen™ TRA-1-60 Alexa Fluor™ 488 Conjugate Kit for Live Cell Imaging > 200 μL of Alexa Fluor™ 488 Mouse Anti-Human TRA-1-60