Learn More
Fisherbrand™ Traceable™ pH/ORP Meter
Designed for water applications, this meter takes the guesswork out of measurements. Fisherbrand™ Traceable™ pH/ORP Meters is ideal for testing pH and ORP in wastewater, water quality and environmental applications.
Brand:  Fisherbrand™ 98769-31
306.86 EUR valid until 2025-04-30
Use promo code "24163" to get your promotional price.
Product Code. 15577583
  • 352.00 EUR / Each
Description
  • Compact instrument fits in the palm of your hand
  • Accuracy: ±0.02 pH
  • 25 point memory storage  with time/date stamp
  • Waterproof (IP67)
  • Triple Display shows pH or ORP measurement, temperature and time\date
  • Ideal for wastewater, water quality and environmental applications
Specifications
Specifications
±20mV
User-selected 1, 2 or 3-point calibration
4277 Traceable™ pH\ORP Meter
pH/ORP Meter
±1°C
0.1º (1º when >100º)
26 x 6.4 x 2.5cm
±0.02
-1800 to 1800 mV
2x CR2032 3V Batteries (included)
0.1mV <1000mV (1mV ≥1000mV)
0 to 60ºC
0 to 60ºC or 32 to 140ºF (selectable)
127 g
Special Offers
  • Multiple Manufacturers
    Up to 60% off
    Promo Code24119
    Expires2025-04-30
  • Fisherbrand
    Get up to 60% off select Fisherbrand products
    Promo Code: 24163
    Expires2025-04-30
    Days
    18
    Hours
    19
    Minutes
    48
    Seconds
    48
    New Offer
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/04/2025 and 30/04/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions Save Now
  • Fisherbrand
    Get up to 60% off select Fisherbrand products
    Promo Code: 24163
    Expires2025-04-30
    Days
    18
    Hours
    19
    Minutes
    48
    Seconds
    48
    New Offer
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/04/2025 and 30/04/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions Save Now
Product Content Correction
Product Title
Fisherbrand™ Traceable™ pH/ORP Meter > pH/ORP Meter