Learn More
Thermo Scientific™ TraceGOLD™ TG-5MS Guard GC Columns with SafeGuard
Extend column lifetime and get more consistent results over a longer period of time.
Brand: Thermo Scientific™ 26098-1305
534.44 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Extend column lifetime and get more consistent results over a longer period of time. Thermo Scientific™ TraceGOLD TG-5MS Guard GC Columns with SafeGuard offer the benefit of an integrated SafeGuard to protect the column.
- With integrated SafeGUARD
- Available in a range of phases
Specifications
330/350°C (Standard Columns), 430°C (Metal Columns) | |
0.25 mm | |
G27, G36 | |
0.25 μm | |
0.25 μm | |
Low-Polar |
0.25 mm | |
TraceGOLD™ TG-5 MS | |
Active Compounds Analysis, GC/MS for Analysis of Active Compounds | |
-60°C to +330/350°C | |
15 m |