missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRANCE/TNFSF11/RANK L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB100-56392
This item is not returnable.
View return policy
Description
TRANCE/TNFSF11/RANK L Polyclonal specifically detects TRANCE/TNFSF11/RANK L in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
TRANCE/TNFSF11/RANK L | |
Polyclonal | |
Unconjugated | |
PBS containing 0.2% gelatin with 0.05% Sodium Azide | |
CD254, CD254 antigen, OPGLOPTB2, Osteoclast differentiation factor, Osteoprotegerin ligand, RANKLreceptor activator of nuclear factor kappa B ligand, Receptor activator of nuclear factor kappa-B ligand, sOdf, TNF-related activation-induced cytokine, TRANCEODFhRANKL2, tumor necrosis factor (ligand) superfamily, member 11, tumor necrosis factor ligand superfamily member 11 | |
Rabbit | |
Protein G purified | |
RUO | |
Primary | |
The amino acid sequence used is 88% homologous in human and 94% homologous in rat. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry | |
0.5 mg/mL | |
Western Blot 0.5 ug/mL, Immunohistochemistry reported in scientific literature (PMID 30082761) | |
NP_003692 | |
TNFSF11 | |
A synthetic peptide corresponding to amino acids 161-177 of mouse CD254 (AQPFAHLTINAASIPSG) was used as immunogen for this antibody. | |
0.1 mg | |
Cancer | |
8600 | |
Human, Mouse, Rat | |
Purified |
For Research Use Only