missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRANCE/TNFSF11/RANK L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
206.95 EUR - 350.96 EUR
Specifications
Antigen | TRANCE/TNFSF11/RANK L |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 0.5 ug/mL, Immunohistochemistry reported in scientific literature (PMID 30082761) |
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18077575
|
Bio-Techne
NB100-56392SS |
0.025 mg |
206.95 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18729653
|
Novus Biologicals
NB100-56392 |
0.1 mg |
350.96 EUR
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRANCE/TNFSF11/RANK L Polyclonal specifically detects TRANCE/TNFSF11/RANK L in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
TRANCE/TNFSF11/RANK L | |
Western Blot 0.5 ug/mL, Immunohistochemistry reported in scientific literature (PMID 30082761) | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
NP_003692 | |
8600 | |
A synthetic peptide corresponding to amino acids 161-177 of mouse CD254 (AQPFAHLTINAASIPSG) was used as immunogen for this antibody. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
0.5 mg/mL | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Cancer | |
PBS containing 0.2% gelatin with 0.05% Sodium Azide | |
CD254, CD254 antigen, OPGLOPTB2, Osteoclast differentiation factor, Osteoprotegerin ligand, RANKLreceptor activator of nuclear factor kappa B ligand, Receptor activator of nuclear factor kappa-B ligand, sOdf, TNF-related activation-induced cytokine, TRANCEODFhRANKL2, tumor necrosis factor (ligand) superfamily, member 11, tumor necrosis factor ligand superfamily member 11 | |
TNFSF11 | |
IgG | |
Protein G purified | |
The amino acid sequence used is 88% homologous in human and 94% homologous in rat. |
For Research Use Only