missing translation for 'onlineSavingsMsg'
Learn More

trans-1-n-Pentyl-4-(4-n-propylcyclohexyl)benzene, 99%, Thermo Scientific™

102.51 EUR - 375.00 EUR

Chemical Identifiers

CAS 89363-50-8
Molecular Formula C20H32
Molecular Weight (g/mol) 272.476
MDL Number MFCD22125193
InChI Key RSUADYMBPNVXJC-UHFFFAOYSA-N
Synonym 1-pentyl-4-trans-4-propylcyclohexyl benzene, 1-4-propylcyclohexyl-4-pentylbenzene, 4-trans-4-propylcyclohexyl pentylbenzene, 4-trans-4-propylcyclohexyl-pentylbenzene, 1-pentyl-4-4-propylcyclohexyl benzene, trans-1-n-pentyl-4-4-n-propylcyclohexyl benzene, 1-pentyl-4-1s,4r-4-propylcyclohexyl benzeneShow MoreShow Less
PubChem CID 15606764
IUPAC Name 1-pentyl-4-(4-propylcyclohexyl)benzene
SMILES CCCCCC1=CC=C(C=C1)C2CCC(CC2)CCC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15442696
SDS View Documents
Thermo Scientific Alfa Aesar
H56443.03
1 g N/A N/A N/A
15452696
SDS View Documents
Thermo Scientific Alfa Aesar
H56443.06
5 g N/A N/A N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
trans-1-n-Pentyl-4-(4-n-propylcyclohexyl)benzene, 99%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.