missing translation for 'onlineSavingsMsg'
Learn More

trans-2-(Benzylamino)cyclohexanol, 99%, Thermo Scientific™

94.87 EUR - 390.00 EUR

Chemical Identifiers

CAS 40571-86-6
Molecular Formula C13H19NO
Molecular Weight (g/mol) 205.301
MDL Number MFCD00145421
InChI Key NJNCYFUGUYIMEQ-CHWSQXEVSA-N
Synonym 1r,2r-2-benzylamino cyclohexanol, trans-2-benzylamino cyclohexanol, trans-2-benzylamino-1-cyclohexanol, 1r,2r-2-benzylamino cyclohexan-1-ol, 1r,2r-2-benzylamino-1-cyclohexanol, cyclohexanol, 2-phenylmethyl amino-, 1r-trans, 2-benzylamino-cyclohexanol, trans-2-benzylamino-cyclohexanol, pubchem19547, trans-2-benzylaminocyclohexan-1-ol
PubChem CID 2724650
IUPAC Name (1R,2R)-2-(benzylamino)cyclohexan-1-ol
SMILES C1CCC(C(C1)NCC2=CC=CC=C2)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11459380
View Documents
Thermo Scientific Alfa Aesar
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11469380
View Documents
Thermo Scientific Alfa Aesar
H50253.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

40571-86-6
205.301
NJNCYFUGUYIMEQ-CHWSQXEVSA-N
2724650
C1CCC(C(C1)NCC2=CC=CC=C2)O
C13H19NO
MFCD00145421
1r,2r-2-benzylamino cyclohexanol, trans-2-benzylamino cyclohexanol, trans-2-benzylamino-1-cyclohexanol, 1r,2r-2-benzylamino cyclohexan-1-ol, 1r,2r-2-benzylamino-1-cyclohexanol, cyclohexanol, 2-phenylmethyl amino-, 1r-trans, 2-benzylamino-cyclohexanol, trans-2-benzylamino-cyclohexanol, pubchem19547, trans-2-benzylaminocyclohexan-1-ol
(1R,2R)-2-(benzylamino)cyclohexan-1-ol

Specifications

40571-86-6
C13H19NO
1r,2r-2-benzylamino cyclohexanol, trans-2-benzylamino cyclohexanol, trans-2-benzylamino-1-cyclohexanol, 1r,2r-2-benzylamino cyclohexan-1-ol, 1r,2r-2-benzylamino-1-cyclohexanol, cyclohexanol, 2-phenylmethyl amino-, 1r-trans, 2-benzylamino-cyclohexanol, trans-2-benzylamino-cyclohexanol, pubchem19547, trans-2-benzylaminocyclohexan-1-ol
C1CCC(C(C1)NCC2=CC=CC=C2)O
205.301
205.3
trans-2-(Benzylamino)cyclohexanol
67°C to 70°C
MFCD00145421
NJNCYFUGUYIMEQ-CHWSQXEVSA-N
(1R,2R)-2-(benzylamino)cyclohexan-1-ol
2724650
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.