missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals trans-L-3-Hydroxyproline

Product Code. 17281306
500 mg
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4298-08-2
Molecular Formula C5H9NO3
Molecular Weight (g/mol) 131.13
MDL Number MFCD00216471
InChI Key BJBUEDPLEOHJGE-IMJSIDKUSA-N
IUPAC Name (2S,3S)-3-hydroxypyrrolidine-2-carboxylic acid
SMILES O[C@H]1CCN[C@@H]1C(O)=O

Specifications

Melting Point 235.0°C
Specific Rotation -17.50
Quantity 500 mg
Formula Weight 131.13
Chemical Name or Material trans-L-3-Hydroxyproline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals trans-L-3-Hydroxyproline >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.