Learn More
Eppendorf™ Transfer Rack for Eppendorf SmartBlock™
Transfer rack suitable for individually sensor-controlled SmartBlock™ heating/cooling/mixing blocks.
Brand: Eppendorf™ 3880000305
This item is not returnable.
View return policy
Description
- Holds up to 24 microtubes for easy transfer of multiple vessels in parallel.
- Convenient transfer of up to 24 hot vessels without burning the fingers
- Temperature range from -86°C to 121°C for broad flexibility
- Limited to ThermoMixer C annd ThermoStat C
Specifications
24 x 0.5 mL | |
SmartBlock, ThermoMixer | |
−86°C to 121 °C |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Eppendorf™ Transfer Rack for Eppendorf SmartBlock™ > Holds 24 x 0.5 mL
Spot an opportunity for improvement?Share a Content Correction