Learn More
Eppendorf™ Transfer Rack for Eppendorf SmartBlock™
Transfer rack suitable for individually sensor-controlled SmartBlock™ heating/cooling/mixing blocks.
Brand:  Eppendorf™ 3880000151
 View more versions of this product
Product Code. 15541290
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
  • Holds up to 24 microtubes for easy transfer of multiple vessels in parallel.
  • Convenient transfer of up to 24 hot vessels without burning the fingers
  • Temperature range from -86°C to 121°C for broad flexibility
  • Limited to ThermoMixer C annd ThermoStat C
TRUSTED_SUSTAINABILITY
Specifications
Specifications
24 x 1.5/2.0 mL
SmartBlock, ThermoMixer
−86°C to 121 °C
Documents
One moment while we fetch your results.
Product Title
Eppendorf™ Transfer Rack for Eppendorf SmartBlock™ > Holds 24 x 1.5/2.0 mL