missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethylhexamethylene Diisocyanate, MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
Specifications
Specifications
CAS | 28679-16-5 |
Quantity | 100 g |
Chemical Name or Material | Trimethyl Hexamethylene Diisocyanate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Trimethylhexamethylene Diisocyanate, MP Biomedicals >
Spot an opportunity for improvement?Share a Content Correction