missing translation for 'onlineSavingsMsg'
Learn More
Tritolyl Phosphate, 63-85%, Technical, Residue other Tolyl Phosphate Isomers, Thermo Scientific Chemicals Product Code.: 10687391

Tritolyl Phosphate, 63-85%, Technical, Residue other Tolyl Phosphate Isomers, Thermo Scientific Chemicals

Product Code. 10687391
500 g, Glass bottle
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10687391

Brand: Thermo Scientific Acros 422315000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1330-78-5
Molecular Formula C21H21O4P
Molecular Weight (g/mol) 368.36
MDL Number MFCD02102116
Synonym Tricresyl phosphate

Specifications

Melting Point -30°C
Color White
Density 1.1600g/mL
Boiling Point 265°C (10.0 mmHg)
Flash Point 234°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (sum of isomers) (GC)
Packaging Glass bottle
Linear Formula (CH3C6H4O)3PO
Refractive Index 1.5550 to 1.557
Quantity 500 g
Merck Index 15, 9940
Specific Gravity 1.16
Solubility Information Solubility in water: insoluble
Formula Weight 368.36
Percent Purity 99%
Grade Technical
Physical Form Powder
Chemical Name or Material Tritolyl phosphate, tech., 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467903 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number422310250,422315000,422310025
Lot NumberA0464779 Certificate TypeCertificate of Analysis Date19/07/2024 Catalog Number422310250,422315000,422310025
Lot NumberA0464777 Certificate TypeCertificate of Analysis Date19/07/2024 Catalog Number422310250,422315000,422310025
Lot NumberA0463102 Certificate TypeCertificate of Analysis Date27/05/2024 Catalog Number422310250,422315000,422310025
Lot NumberA0450445 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number422310250,422315000,422310025
compliance-icons compliance-icons
Product Identifier
  • Tritolyl phosphate, mixture of isomers
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Reproductive toxicity Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P391-Collect spillage.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tritolyl Phosphate, 63-85%, Technical, Residue other Tolyl Phosphate Isomers, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.