Learn More
Trovirdine, MedChemExpress
MedChemExpress Trovirdine inhibits HIV-1 RT with an IC50 of 7 nM when employing heteropolymeric primer/template (oligo-DNA/ribosomal RNA)and dGTP as substrate.
Brand: Medchem Express HY-15349
Description
Trovirdine inhibits HIV-1 RT with an IC50 of 7 nM when employing heteropolymeric primer/template (oligo-DNA/ribosomal RNA)and dGTP as substrate.Specifications
Trovirdine | |
White | |
C13H13BrN4S | |
LY300046 | |
S=C(NCCC1=NC=CC=C1)NC2=NC=C(Br)C=C2 | |
337.24 | |
Research | |
COVID-19-anti-virus |
149488-17-5 | |
Research | |
10 mg | |
DMSO : 100 mg/mL (296.52 mM; Need ultrasonic) | |
337.24 | |
98.59% | |
Solid |