Learn More
Thermo Scientific™ TSG General Purpose Refrigerators - Boxed
The TSG Series of general purpose refrigerators are built with the performance, security, and quality features you have come to expect from Thermo Scientific™.
Brand: Thermo Scientific™ TSG400REGBV
Description
The Thermo Scientific™ TSG Series general purpose laboratory refrigerators are designed with your needs in mind. The TSG series is ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing reagents, media, proteins, and samples used daily in the lab.
TSG Series General Purpose Refrigerators are designed with your laboratory in mind, combining the performance, security, and quality required for everyday laboratory applications.
- Wide range of sizes from 400 L to 1500 L double door
- Glass door (triple pane) and solid door configurations
- Whisper quite operation < 50 dBA
- White painted interior and exterior
- Adaptive Defrost
- Temperature, door ajar, and power failure (with battery backup) alarms.
- Low heat output
- Rear access port
- Keyed Lock
- LED interior lighting
- Green hydrocarbon refrigerants
- Remote alarm contacts
Specifications
TSG Series General Purpose Laboratory Refrigerator | |
2°C to 10°C | |
R290 | |
1 | |
30 kg | |
Painted Steel | |
CE | |
Digital Microprocessor | |
36 months | |
Boxed | |
Key Lock | |
493 x 500 x 1405 mm | |
EU | |
230 V/50 Hz | |
Energy Efficient |
400 L | |
+5°C | |
Glass | |
5 Adjustable | |
Steel | |
Painted Steel | |
Yes | |
1 Rear Access Port | |
Adaptive Defrost | |
Alarm | |
Smart-Vue Pro Compatible | |
650 x 600 x 1955 mm | |
230 V | |
50 Hz | |
150 kg |