Learn More
Tungsten Standard Metal Solution 1000ppm, for ICP, Solution NH3 0.1M, Fisher Chemical
Click to view available options
Quantity:
500 mL
Specifications
Specifications
Packaging | HDPE plastic bottle |
Quantity | 500 mL |
MDL Number | 11461 |
Physical Form | Liquid |
Color | Colorless |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 6
Safety and Handling
Product Identifier
- Tungsten solution 1000 ppm
Product Content Correction
Product Title
Tungsten Standard Metal Solution 1000ppm, for ICP, Solution NH3 0.1M, Fisher Chemical >
Spot an opportunity for improvement?Share a Content Correction