Learn More
Thermo Scientific™ TX-200 Rotor Tube Adapters
Use Thermo Scientific™ TX-200 Rotor Tube Adapters with round buckets in the TX-200 Rotor.
Brand: Thermo Scientific™ 75003810
This item is not returnable.
View return policy
Description
Use Thermo Scientific™ TX-200 Rotor Tube Adapters with round buckets in the TX-200 Rotor.
Adapters hold a variety of tubes including round-bottom, conical, urine, blood collection and microtubes.- Maximum tube capacity: 4 × 100mL
- Maximum O.D. × L: 48 × 122mm
Specifications
Rotor Tube Adapter | |
5/7 mL Round-bottom Tube | |
13 mm | |
118 mm |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 6
4. GP_Multifuge X1_X4 Pro_Megafuge ST1_ST4 Plus_EN_1223.pdf
File size: 3.3 MB
Special Offers
-
Multiple Manufacturers
The Academic Best Buy Book 2025
Browse exclusive prices for our academic customersPromo Code: 25081Expires: 2025-12-31
Product Content Correction
Product Title
Thermo Scientific™ TX-200 Rotor Tube Adapters > 5/7mL Round-bottom tube
Spot an opportunity for improvement?Share a Content Correction