Learn More
DuPont™ Tychem™ 6000F Gown Product Code.: 16608525
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
DuPont™ Tychem™ 6000F Gown
Product Code. 16608525 Shop All DuPont Products
Medium
Click to view available options
Size:
Medium
2X-Large
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 16608525
Brand: DuPont™ D15546624
Estimated Shipment: 29-04-2025
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Shin-length gown with wrap-over rear closure, hook and loop neck closure and waist ties. Double cuffs for increased protection and tighter connection with gloves.
  • Elasticated wrists
  • Knitted cuff (inside) for better comfort
  • Mandarin collar for improved neck and throat area closure
  • Offers excellent chemical permeation protection to an extensive range of chemicals, helping to protect personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, biohazards and certain chemical warfare agents.
TRUSTED_SUSTAINABILITY
Specifications
Size Medium
Color White
Cuff Style Elasticated
Product Content Correction
Product Title
DuPont™ Tychem™ 6000F Gown >