missing translation for 'onlineSavingsMsg'
Learn More

Saint Gobain Life Sciences™ Tygon™ E-3603 Laboratory tubing

E-3603 Tubing for laboratory and vacuum applications.

Brand:  Saint Gobain Life Sciences™ ACFUN030

 View more versions of this product

Product Code. 17933821

  • 867.00 EUR / 10m

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Crystal clear and flexible, handling virtually all organic chemicals found in the modern laboratory
  • Non-oxidizing and non-contaminating, making it an ideal choice for a variety of applications
  • High tolerances and lot-to-lot consistency, which will yield reproducible results in your laboratory
  • Less permeable than rubber tubing
  • Glassy-smooth inner bore helps prevent buildup to facilitate cleaning
  • Specially formulated for resistance to flex-fatigue and abrasion
  • Tubing for instrumentation connection, vent, drain and other general laboratory applications
  • Offers long life, which minimizes labor and expenses associated with replacement
  • Lot-to-lot consistency for reproducible results
  • Smooth, polished inner wall
  • Slips easily over fittings and grips securely for simple lab set-ups
  • Contains no BPA or phthalates
  • Standard sizes available to hold full vacuum at room temperature
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Autoclavable
9.5 mm
0.878 in.
32.8 in.
Laboratory and Vacuum Applications
Clear
Flexible
165°F
Tubing
0.374 in.
Thermoplastic
22.3 mm
10 m
173 lbf/in.
56 (Shore A)
51 psi
74°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.