Learn More
Ismatec™ Tygon™ E-Lab C/L™ Pump Tubing
Ideal for general transfer applications
Brand:  Ismatec™ 06460-38
 View more versions of this product
Product Code. 15480305
  • 185.00 EUR / 30m
Estimated Shipment: 10-03-2025
to see stock.
Explore more special offers
Description
  • Tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
  • For general laboratory use
  • Nontoxic, non-aging, non-oxidizing, easy flow monitoring, transparent
  • The overall length of E-Lab 06460-series extension tubing is 100 feet (30 m). The legacy Ismatec extension tubing was 10 m long.
Specifications
Specifications
Prop 65 NA (no Prop 65 chemicals)
Autoclavable
Tygon E-Lab (E-3603)
30.4 m
38
25.3 psi, 1.75 bar
Microbore Tubing
1.65 mm
100 ft.
0°C
1/16 in.
40°C