Learn More
Masterflex™ Tygon™ Lab Tubing
Formulated for resistance to flex fatigue and abrasion and is less permeable than rubber tubing. Saint Gobain Performance Plastics™ Tygon™ Lab Tubing handles virtually all inorganic chemicals found in the lab.
Brand:  Masterflex™ 06407-96
 View more versions of this product
Product Code. 15446633
  • 733.00 EUR / 50ft
Estimated Shipment: 04-06-2025
to see stock.
Explore more special offers
Description
  • Alternative to DEHP tubing
  • New formulation delivers the same excellent performance
  • Contains no BPA
  • Outstanding chemical resistance
  • Lot-to-lot consistency for reproducible results
  • Increases productivity in peristaltic pumps – outlasts other clear tubing 2 to 1
  • Smooth, polished inner bore
  • Non-oxidizing and non-contaminating
  • Slips easily over fittings and grips securely
  • Coils marked at 1-foot intervals for easy measuring
Specifications
Specifications
FDA compliant; meets USP Class VI and NSF 51 requirements
19.1 mm
25.4 mm
3.2 mm
-46°C
Sterile
14 psi
Autoclavable
Tygon E-3603
15.24 m
General laboratory, food and beverage, biopharmaceutical, analytical instruments
1 Pack
Transparent
74°C