Learn More
TZ9, MedChemExpress
MedChemExpress TZ9 is a novel inhibitor of Rad6 ubiquitin conjugating enzyme(E2 enzyme); inhibits MDA-MB-231 cell proliferation with IC50 of ∼6 uM.

139.05 EUR - 781.65 EUR
Specifications
Chemical Name or Material | TZ9 |
---|---|
Physical Form | Solid |
CAS | 1002789-86-7 |
Color | White |
Purity Grade Notes | Research |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16490028
![]() |
Medchem Express
HY-18643 |
5 mg |
139.05 EUR
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16453577
![]() |
Medchem Express
HY-18643 |
50 mg |
781.65 EUR
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16447558
![]() |
Medchem Express
HY-18643 |
10 mg |
222.75 EUR
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TZ9 is a novel inhibitor of Rad6 ubiquitin conjugating enzyme(E2 enzyme)Specifications
TZ9 | |
1002789-86-7 | |
Research | |
Cancer-programmed cell death | |
O=C(OCC1=NC(N)=NC(NC2=CC=CC=C2)=N1)C3=CC=C([N+]([O-])=O)C=C3 | |
366.33 | |
Research |
Solid | |
White | |
C17H14N6O4 | |
DMSO : ≥ 40 mg/mL (109.19 mM) | |
366.33 | |
99.26% |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.