Learn More
uvex Unigrip 6624 Cotton/Polyamide Assembly Gloves with PVC Dot Palm
Shop All Uvex Products
Click to view available options
Size:
7
8
9
Description
- Gloves are constructed of a 10 gauge cotton/polyamide blend
- PVC dots on the palm enhance grip capabilities in dry applications
- Ideal for assembly, sorting, and course mechanical work
Specifications
Specifications
Coating Material | PVC Dots |
Color | Gray, Red |
Disposable | Reusable |
Exterior Finish | Smooth |
Hand | Ambidextrous |
Material | Cotton, Polyamide |
Powdered | Powder Free |
Product Type | Work Gloves |
Size | 9 |
Sterility | Non-sterile |
Show More |
Product Content Correction
Product Title
uvex Unigrip 6624 Cotton/Polyamide Assembly Gloves with PVC Dot Palm >
Spot an opportunity for improvement?Share a Content Correction