Learn More
Cole-Parmer™ Union Threaded Elbow
Threaded fittings offer more permanent installation
Brand:  Cole-Parmer™ 06546-34
 View more versions of this product
Product Code. 11868033
  • 117.00 EUR / Each
Estimated Shipment: 17-02-2025
to see stock.
Explore more special offers
Description
  • Chemically-inert PFA handles most aggressive chemicals
  • Threaded reducing bushing
  • Choose the size and material that meets your application needs
  • Use with our rigid tubing for your high-pressure applications
Specifications
Specifications
Union Threaded Elbow
Not Autoclavable
3/8 in.
3/8 in. NPTF x 3/8 in. NPTF
NPT(F) Connection
PFA
Ethylene Oxide
Translucent