missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Urea, ≥99%, (Crystalline/ACS Reagent Grade), MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 kg
5 kg
Specifications
Specifications
Assay Percent Range | ≥99% |
Quantity | 1 kg |
Formula Weight | 60.06 |
Grade | ACS Reagent |
Physical Form | Crystalline |
Chemical Name or Material | Urea |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Urea, ≥99%, (Crystalline/ACS Reagent Grade), MP Biomedicals >
Spot an opportunity for improvement?Share a Content Correction