Learn More
Hirschmann™ USP Calibration Certificate
Individual calibration certificate for USP glassware with an individual serial number ID on the instrument and the certificate.
Brand: Hirschmann™ 9980104
This item is not returnable.
View return policy
Description
- Individual USP calibration certificate
- Markings with instrument ID
- The quality certificate and the instrument are equipped with an individual serial number
- The certificate provides the following information: the nominal volume, the prescribed tolerance according USP, the actually determined volume of the instrument and the uncertainty of measurement
Specifications
Volumetric Glassware, Class A/AS, Confomity Certified acc. to USP | |
USP Certificate of Performance |
Product Content Correction
Product Title
Hirschmann™ USP Calibration Certificate > For Use With (Equipment): Volumetric glassware, Class A/AS, confomity certified acc. to USP
Spot an opportunity for improvement?Share a Content Correction