Learn More
uvex C500 Foam Cut Protection Gloves
Shop All Uvex Products
Click to view available options
Size:
6
7
8
9
10
11
Description
Specifications
Specifications
Certifications/Compliance | EN 388:2016 (4 X 4 2 C), 407 (X 1 X X X X), 388 (4 5 4 2) |
Coated Area | Finger tips and palm |
Description | Bamboo TwinFlex™, uvex™ 3D Ergo, and uvex™ climazone hand climate management technologies |
Coating Material | HPE |
Color | Lime Green, Anthracite |
Cut Resistance Rating | ANSI A5 |
Hand | Ambidextrous |
Latex Free | Yes |
Material | HPPE, Polyamide |
Product Type | Cut-resistant Gloves |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 10
Special Offers
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Product Content Correction
Product Title
uvex C500 Foam Cut Protection Gloves >
Spot an opportunity for improvement?Share a Content Correction