Learn More
Thermo Scientific™ Valco Injector, Model C6W
Valco continues to offer a diverse line in terms of number of ports, fitting sizes, and materials of construction.
Brand: Thermo Scientific™ EPC6W
2305.44 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Specifications
Microelectric | |
EPC6 W | |
Injector |
20 μL Loop | |
Six 0.016 in. |