Learn More
R&D Systems™ Mouse FGFR3 (NP_032036) VersaClone cDNA
FGFR3 plays an essential role in the regulation of cell proliferation, differentiation, and apoptosis
Brand:  R&D Systems™ RDC1204
Encompass
Product Code. 15731195
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
The Mouse FGF R3 (NP_032036) VersaClone cDNA clone contains the complete ORF for FGFR3, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The FGFR3 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature
Specifications
Specifications
NP_032036
cDNA is provided in 10mM Tris-Cl, pH 8.5
CD333, CEK, EC 2.7.10, FGF R3, fibroblast growth factor receptor 3, HSFGFR3EX, JTK4, JTK4thanatophoric dwarfism
Mouse
10 μg at 0.2 μg/μL
2416bp
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
10 μg
Documents
One moment while we fetch your results.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
R&D Systems™ Mouse FGFR3 (NP_032036) VersaClone cDNA
For Reasearch Use Only; Not for Use in Humans