Learn More
3M™ Versaflo™ Powered Air Respirator Starter Kit, TR-619E
3M™ Versaflo™ Powered Air Turbo Starter Kit, TR-619E is our ready-to-use kit for respiratory protection against particles, gases and vapours. It comes with everything you need to get started. Simply add your 3M™ Versaflo™ Headtop for complete respiratory protection.
Brand:  3M™ 7100053532
1474.58 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15559255
  • 2180.00 EUR / Each
Description
  • Simple starter kit with all the component parts to get you up and running. Simply add the headtop required
  • Easy to use, versatile respiratory protection against dusts, mists, fumes and organic vapours
  • Kit includes Turbo Unit TR-602E, A2P filter, filter cover, pre-filters, spark arrestors, easy clean belt, high capacity battery, single station battery charger kit, BT-30 length adjusting breathing tube and airflow indicator
  • Compatible headtop required
  • Visual, vibratory and audible warning alarms to alert to low battery and air flow
  • System run time of 9.5-19 hours depending on battery, flow rate, headtop and filter
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PAPR Assembly, No Headtop
Versaflo™
Videos
  • 3M - Versaflo powered air turbo TR-600
    3M Versaflo TR-600
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      3M Versaflo powered air turbo TR-600
    Certificates
    View accessory products
    Product Title
    3M™ Versaflo™ Powered Air Respirator Starter Kit, TR-619E > Product Type: PAPR System Kit, Color: Light Gray, Model: TR-619E