Learn More
Invitrogen™ ViewRNA™ Temperature Validation Kit
The ViewRNA Temperature Validation Kit uses a calibrated digital thermometer to assess temperature accuracy.
Brand: Invitrogen™ QV0523
Description
The ViewRNA Temperature Validation Kit uses a calibrated digital thermometer to assess temperature accuracy. This kit may be used for any ViewRNA Assay to evaluate the temperature of incubators or ThermoBrite systems.
Specifications
Temperature Validation Kit | |
1 Kit |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
441815-000 | Certificate of Analysis | 14/04/2025 | |
440692-000 | Certificate of Analysis | 10/04/2025 | |
435988-000 | Certificate of Analysis | 19/12/2024 | QV0523 |
412973-000 | Certificate of Analysis | 07/06/2024 | QV0523 |
409764-000 | Certificate of Analysis | 09/05/2024 | QV0523 |