Learn More
Emma Safety Footwear Vila Safety Shoes
Lace-up shoe with zipped cover provides additional protection against all kinds of dirt, germs and bacteria. Features SmartecAir™ for dry, fresh feet.
Brand: Emma Safety Footwear MM54654437
This item is not returnable.
View return policy
Description
- S2-Safety shoe for wet working conditions
- Microfiber construction
- Lace-up front with zipper covering
- GripForce™ Easy Twist Outsole
- Chrome-free lining
- Hydro-Tec™ Sanitized Silver
Specifications
EN ISO 20345:2011 | |
White | |
Hydro-Tec™ Sanitized Silver | |
37 |
Laces | |
Vila | |
Microfiber | |
Steel |