Learn More
Emma Safety Footwear Vila Safety Shoes
Lace-up shoe with zipped cover provides additional protection against all kinds of dirt, germs and bacteria. Features SmartecAir™ for dry, fresh feet.
Specifications
Certifications/Compliance | EN ISO 20345:2011 |
---|---|
Closure Type | Laces |
Color | White |
Design | Vila |
Liner Material | Hydro-Tec™ Sanitized Silver |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15978731
|
Emma Safety Footwear
MM54654435 |
35 |
N/A
|
N/A | |||||
15988731
|
Emma Safety Footwear
MM54654436 |
36 |
N/A
|
N/A | |||||
15998731
|
Emma Safety Footwear
MM54654437 |
37 |
N/A
|
N/A | |||||
15908741
|
Emma Safety Footwear
MM54654438 |
38 |
N/A
|
N/A | |||||
15918741
|
Emma Safety Footwear
MM54654439 |
39 |
N/A
|
N/A | |||||
15928741
|
Emma Safety Footwear
MM54654440 |
40 |
N/A
|
N/A | |||||
15938741
|
Emma Safety Footwear
MM54654441 |
41 |
N/A
|
N/A | |||||
15948741
|
Emma Safety Footwear
MM54654442 |
42 |
N/A
|
N/A | |||||
15958741
|
Emma Safety Footwear
MM54654443 |
43 |
N/A
|
N/A | |||||
15968741
|
Emma Safety Footwear
MM54654444 |
44 |
N/A
|
N/A | |||||
15978741
|
Emma Safety Footwear
MM54654445 |
45 |
N/A
|
N/A | |||||
15988741
|
Emma Safety Footwear
MM54654446 |
46 |
N/A
|
N/A | |||||
15998741
|
Emma Safety Footwear
MM54654447 |
47 |
N/A
|
N/A | |||||
15908751
|
Emma Safety Footwear
MM54654448 |
48 |
N/A
|
N/A | |||||
Description
- S2-Safety shoe for wet working conditions
- Microfiber construction
- Lace-up front with zipper covering
- GripForce™ Easy Twist Outsole
- Chrome-free lining
- Hydro-Tec™ Sanitized Silver
Specifications
EN ISO 20345:2011 | |
White | |
Hydro-Tec™ Sanitized Silver | |
Steel |
Laces | |
Vila | |
Microfiber |