missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Vitamin K Control Diet For Rat, Pellet

Contains vitamins and minerals.

Brand:  MP Biomedicals 0296042610

 View more versions of this product

Product Code. 11431522

  • 662.00 EUR / 10kg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Ideal for experiments containing control vs. modified diets.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Autoclavable
Single-use
10 kg
Refrigerated
0.3048 m
Animal Diet
Pellet
Non-sterile
1 ft.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.