Learn More
Water with 0.1% Formic Acid (v/v), Certified HPLC for Gradient Analysis Product Code.: 12317123
Water with 0.1% Formic Acid (v/v), Certified HPLC for Gradient Analysis
Product Code. 12317123
2.5L, Amber glass bottle
Click to view available options
2.5L, Amber glass bottle
Product Code. 12317123
Brand: Fisher Chemical FE/1200/17
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Quantity 2.5 L
Product Content Correction
Product Title
Water with 0.1% Formic Acid (v/v), Certified HPLC for Gradient Analysis >