missing translation for 'onlineSavingsMsg'
Learn More
Water Deionized, Solveco Product Code.: 18032203
Estimated Shipment: 05-05-2025
to see stock.
Add to basket

Water Deionized, Solveco

Product Code. 18032203
25 L
missing translation for 'orderingAttributeHoverText'
Quantity:
5 L
25 L
This item is not returnable. View return policy

Product Code. 18032203

Brand: Solveco Chemicals 1105

Estimated Shipment: 05-05-2025
to see stock.
Add to basket
Add to basket
This item is not returnable. View return policy

Deionized water is highly purified water with most dissolved ions removed through ion-exchange processes. This makes it ideal for sensitive applications.

Deionized water is highly purified water with most dissolved ions removed through an ion-exchange process. This results in water with very low electrical conductivity and minimal impurities. It is commonly used in laboratories, electronics cleaning, and cooling systems where high purity is essential. The lack of ions helps prevent contamination and scaling, ensuring reliable results and extending the lifespan of equipment.

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Percent Purity 99.90%
Grade Technical
Packaging HDPE Container
Sterility Non-sterile
Quantity 25 L
Chemical Name or Material Dihydrogen Monoxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Water Deionized, Solveco >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.