Learn More
Cytiva Whatman™ Regenerated Cellulose Membranes: Grade RC 55
Consists of pure cellulose without any wetting agents.
Brand: Cytiva 10410214
277.01 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
This item is not returnable.
View return policy
Description
- Spontaneously wetting, very good wet strength
- Extremely chemically resistant; suitable for aqueous and organic media
- Hydrophilic
- Mechanically stable
- Sterilizable by all methods
Specifications
Regenerated Cellulose | |
Hydrophilic | |
50 mm |
0.5 μm | |
75 μm |
WHA015 SYNCHRONIZED SOLUTIONS FOR QC
File size: 4 MB
Product Content Correction
Product Title
Cytiva Whatman™ Regenerated Cellulose Membranes: Grade RC 55 > Diameter: 50 mm
Spot an opportunity for improvement?Share a Content Correction