Learn More
DWK Life Sciences Wheaton™ Perforated Aluminum Capsules
GREENER_CHOICE
Tear-off seals remove completely allowing content to pour from bottle. Wheaton™ Tear-Off Aluminum Seals, Unlined are available in a wide range of sizes and colors.
Brand:  DWK Life Sciences 224193-06
113.99 EUR valid until 2025-12-31
Use promo code "24362" to get your promotional price.
 View more versions of this product
Product Code. 10580463
  • 123.00 EUR / Pack of 1000
Estimated Shipment: 24-04-2025
Description
Tear-off seals remove completely allowing content to pour from bottle. Wheaton™ Tear-Off Aluminum Seals, Unlined are available in a wide range of sizes and colors.
  • Good barrier properties
  • Color selection includes natural, red, blue and green
  • Chemically resistant to alcohol and acetone, among many others
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Red
1000/Cs.
20 mm
Aluminum
Complete Tear-off Cap
SDS
Documents Certificates
Special Offers
Special Offers
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
    Days
    21
    Hours
    14
    Minutes
    2
    Seconds
    34
Product Content Correction
Product Title
DWK Life Sciences Wheaton™ Perforated Aluminum Capsules > Complete Tear-Off; Red; For mouth O.D.: 20mm; 1000/Cs.