Learn More
DWK Life Sciences Wheaton™ BOD Bottle Cap
For use with Wheaton 60 and 300mL BOD bottles and 300mL Black Borosilicate BOD bottles
Brand:  DWK Life Sciences 227723
79.94 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Product Code. 10572744
  • 84.15 EUR / Pack of 50
Estimated Shipment: 30-04-2025
Description
For use with Wheaton 60 and 300mL BOD bottles and 300mL Black Borosilicate BOD bottles
  • Polyethylene cap prevents evaporation of water seal during incubation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Prevents evaporation of the water seal during the five-day BOD incubation period
For 60 and 300mL BOD bottle
Polyethylene
Product Title
DWK Life Sciences Wheaton™ BOD Bottle Cap > Bottle cap; For 60 and 300mL B.O.D. bottles