Learn More
DWK Life Sciences Wheaton™ E-Z Decappers
With ergonomically designed handles for sealing and decapping standard laboratory vials
Brand: DWK Life Sciences W225351
This item is not returnable.
View return policy
Description
Wheaton E-Z Decappers are recommended for decapping standard laboratory vials.
- Several sizes of decapper are available, including those that remove seals 8mm, 11mm, 13mm and 20mm wide.
- The decappers come with visible size labels for easy identification.
- User comfort is preserved by an ergonomically designed, cushioned handle. Hand fatigue is reduced by the short handle and redesigned spring assembly.
- Decappers can be autoclaved, although autoclaving may change the appearance or reduce product life. They should not be autoclaved at greater than 121°C
Services
Two years
Specifications
11 mm Seals |
Product Suggestions
Customers who viewed this item also viewed
WHE026 Catalogue 2012 FINAL LOWRES
File size: 14.8 MB
WHE026 Catalogue 2012 FINAL LOWRES
File size: 14.8 MB
Accessories and Services
Optional Accessory
-
DWK Life Sciences Wheaton™ Crimpers E-Z Crimper, Standard Seal 11mm Product Code: 10670884
539.00 EUR / Each
Product Content Correction
Product Title
DWK Life Sciences Wheaton™ E-Z Decappers > Seal Decapper; 11mm Seals
Spot an opportunity for improvement?Share a Content Correction