Learn More
Whirl-Pak™ Polyethylene Write-on Bags
GREENER_CHOICE
Brand:  Whirl Pak B01515WA
212.21 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
 View more versions of this product
Product Code. 11744834
  • 295.00 EUR / Pack of 500
Estimated Shipment: 04-06-2025
Specifications
2041 mL
Special Offers
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
    Days
    0
    Hours
    15
    Minutes
    51
    Seconds
    40
Product Content Correction
Product Title
Whirl-Pak™ Polyethylene Write-on Bags > Capacity: 2041mL