missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Wide Neck PTFE Bottle

Product Code. 10685131 Shop All Brand Products
250 mL
Click to view available options
Capacity (Metric):
250 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Outstanding chemical and thermal resistance

  • Sturdy, strong-walled design
  • Includes screw cap
  • TRUSTED_SUSTAINABILITY

    Specifications

    Capacity (Metric) 250 mL
    Height (Metric) 120 mm
    Diameter (Metric) 67 mm
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    BRAND™ Wide Neck PTFE Bottle >

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.