Learn More
Micronova™ Wipe Controlled Environment
Brand: Micronova™ WPAA-1218
This item is not returnable.
View return policy
Description
single ply, over-locked edge, microfibre, 305mm x 457mmProduct Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
040316-000 | Certificate of Compliance | 12/10/2023 | WPAA-1218 |
037066-000 | Certificate of Compliance | 22/03/2023 | WPAA-1218 |
032282-000 | Certificate of Compliance | 16/05/2022 | WPAA-1218 |
028527-000 | Certificate of Compliance | 13/10/2021 | WPAA-1218 |
026102-000 | Certificate of Compliance | 28/05/2021 | WPAA-1218 |
Product Content Correction
Product Title
Micronova™ Wipe Controlled Environment >
Spot an opportunity for improvement?Share a Content Correction