Learn More
Abeba™ X-Light Safety Loafer
Abeba X-Light Black Safety Loafer - Smooth leather safety shoes with elastic instep.
Brand:  Abeba™ 711037/37
 View more versions of this product
Product Code. 17631915
  • 106.00 EUR / 1 pair
Estimated Shipment: 12-05-2025
to see stock.
Explore more special offers
Description
  • CE, EN ISO 20345:2011, S2, SRC
  • Smooth leather
  • Black
  • Steel toe cap
  • Slip resistance PU outsole sole
  • Replaceable Active Comfort Insole (art. 351620)
  • Antistatic
  • Breathable lining with Silver ions
  • Instep area with elastic band
Specifications
Specifications
Elastic Band
Leather
Steel
Black
37
CE, EN ISO 20345:2011, S2, SRC
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Abeba™ X-Light Safety Loafer > Safety Loafer; Size: 37