Learn More
Abeba™ X-Light Safety Loafer
Abeba X-Light Black Safety Loafer - Smooth leather safety shoes with elastic instep.
Brand: Abeba™ 711037/47
This item is not returnable.
View return policy
Description
- CE, EN ISO 20345:2011, S2, SRC
- Smooth leather
- Black
- Steel toe cap
- Slip resistance PU outsole sole
- Replaceable Active Comfort Insole (art. 351620)
- Antistatic
- Breathable lining with Silver ions
- Instep area with elastic band
Specifications
Elastic Band | |
Leather | |
Steel |
Black | |
47 | |
CE, EN ISO 20345:2011, S2, SRC |
Product Content Correction
Product Title
Abeba™ X-Light Safety Loafer > Safety Loafer; Size: 47
Spot an opportunity for improvement?Share a Content Correction