Learn More
Lonza X-VIVO™ 15
Optimized for the proliferation of Tumor Infiltrating Lymphocytes (TIL) under serum-free conditions. X-VIVO™ 15 is a chemically defined, serum-free hematopoietic cell medium.
Brand:  Lonza BE02-060Q
Product Code. 11695120
Description
Description
Formulated with:
  • L-glutamine
  • gentamicin
  • phenol red
Order Info
dimProductType
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cell Culture Media
Product Title
Lonza X-VIVO™ 15 > Product Type: Culture Media