Learn More
ZEOprep™ SH (Thiol) Functionalized
ZEOprep™ is a packing material providing fast, effective, and reproducible separations. THIOL Irregular shaped silica gel is used in Gravity, Flash, and Medium Pressure Chromatography. The THIOL ligand makes an Ion Exchange material suitable for weak anion interactions.
Brand:  ZEOprep 302083
 View more versions of this product
Product Code. 30049005
  • 938.00 EUR / 500g
Description
  • SH (Thiol) functionalized for Ion Exchange separations
  • Idea for weak anion applications
  • Irregular Amorphous Silica Gel
  • High Purity
  • European Manufacture
  • Swiss accuracy and consistency
  • Excellent batch to batch reproducibility
  • ISO Certification
  • High Chemical and Mechanical Stability
  • Narrow Particles Size Distribution
  • 20 to 500 μm particles size range
  • Available in 60 A or 90 A pore size
  • High loadability
  • Selection of pack sizes available
  • Suitable for gravity, flash, prep and process scale
Specifications
Specifications
40 to 63 μm
Chromatography
Silica Gel
SH (Thiol) Functionalized
60 Å
500 g
Silica Gel
Irregular
Product Title
ZEOprep™ SH (Thiol) Functionalized > 500 g