missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Sulfate Heptahydrate, ACS Reagent Grade, Scharlab
Click to view available options
Quantity:
500 g
Description
- Synonyms: Sulfuric acid zinc salt heptahydrate, Zinc vitriol
- Molecular Formula: ZnSO4 • 7H2O
- Molar Mass: 287.54g/mol
- CAS: 7446-20-0
- EIN ECS No.: 231-793-3
Specifications
Specifications
Melting Point | ∼40°C (decomposition) |
CAS | 7446-20-0 |
pH | 4.4–6.0 (5%, water) |
Quantity | 500 g |
Molecular Formula | H14O11SZn |
MDL Number | MFCD00149894 |
Synonym | zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o |
InChI Key | RZLVQBNCHSJZPX-UHFFFAOYSA-L |
SMILES | O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O |
IUPAC Name | zinc(2+) heptahydrate sulfate |
Show More |
Safety and Handling



Product Identifier
- Zinc sulfate heptahydrate, reagent grade, ACS, ISO, Reag. Ph Eur
- Danger
- Serious eye damage/eye irritation Category 1
- Acute toxicity Category 4
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P273-Avoid release to the environment.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P501-Dispose of contents/container to .
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Zinc Sulfate Heptahydrate, ACS Reagent Grade, Scharlab >
Spot an opportunity for improvement?Share a Content Correction